Abnova
Manufacturer Code:PAB22149
Catalog # 89-123-856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human DEM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant DEM1.
For Research Use Only
Protein Aliases: C1orf176 FLJ11445 FLJ13183 FLJ21144; defects in morphology 1 homolog; Defects in morphology protein 1 homolog; Exo V; Exonuclease V; probable exonuclease V
Gene Aliases: C1orf176; DEM1; Exo V; EXO5; hExo5
UniProt ID: (Human) Q8WTW9
Entrez Gene ID: (Human) 64789
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.