Proteintech
Manufacturer Code:164481AP150UL
Catalog # 50-173-0950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DEK Fusion Protein Ag8225 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D6S231E DEK DEK oncogene Protein DEK; DEK oncogene (DNA binding); Protein DEK
Gene Aliases: 1810019E15Rik; D13H6S231E; D6S231E; DEK
UniProt ID: (Human) P35659, (Rat) Q6AXS3, (Mouse) Q7TNV0
Entrez Gene ID: (Human) 7913, (Rat) 306817, (Mouse) 110052
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.