Invitrogen
Manufacturer Code:PA524082
Catalog # PIPA524082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 261-290 amino acids from the C-terminal region of human DEGS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf66 DES2 FADS8 degenerative spermatocyte homolog 2 lipid desaturase sphingolipid C4-hydroxylase/delta 4-desaturase sphingolipid delta 4 desaturase/C-4 hydroxylase sphingolipid delta(4)-desaturase 2 sphingolipid delta(4)-desaturase/C4-hydroxylase DES2; Degenerative spermatocyte homolog 2; degenerative spermatocyte homolog 2, lipid desaturase; DEGS2; delta(4)-desaturase, sphingolipid 2; dihydroceramide desaturase 2; Sphingolipid 4-desaturase; sphingolipid C4-hydroxylase/delta 4-desaturase; Sphingolipid C4-monooxygenase; sphingolipid delta 4 desaturase/C-4 hydroxylase; sphingolipid delta(4)-desaturase 2; sphingolipid delta(4)-desaturase/C4-hydroxylase DES2; Sphingolipid delta(4)-desaturase/C4-monooxygenase DES2
Gene Aliases: C14orf66; DEGS2; DES2; FADS8
UniProt ID: (Human) Q6QHC5
Entrez Gene ID: (Human) 123099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.