Invitrogen
Manufacturer Code:PA542741
Catalog # PIPA542741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DEGS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; Degenerative spermatocyte-like protein RDES; Delta(4)-desaturase sphingolipid 1 DEGS DES1 Des-1 FADS7 MGC5079 MIG15 MLD DEGS-1; delta(4)-desaturase, sphingolipid 1; dihydroceramide desaturase; dihydroceramide desaturase 1; Dihydroceramide desaturase-1; membrane fatty acid (lipid) desaturase; Membrane lipid desaturase; migration-inducing gene 15 protein; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; Sphingolipid delta(4)-desaturase DES1
Gene Aliases: AA536663; DEGS; DEGS-1; DEGS1; Des-1; DES1; FADS7; fb81d06; im:6909319; Mdes; MIG15; MLD; wu:fa25h01; wu:fb81d06
UniProt ID: (Bovine) Q3ZBY7, (Human) O15121, (Mouse) O09005, (Rat) Q5XIF5
Entrez Gene ID: (Horse) 100055442, (Sheep) 100192315, (Bovine) 507290, (Goat) 102181802, (Rabbit) 100355393, (Human) 8560, (Mouse) 13244, (Guinea pig) 100717671, (Rat) 58970, (Zebrafish) 327075
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.