Abnova
Manufacturer Code:PAB9854
Catalog # 89-134-456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human DEGS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DEGS1.
For Research Use Only
Protein Aliases: Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; DEGS; delta(4)-desaturase, sphingolipid 1; Des-1; DES1; dihydroceramide desaturase 1; Dihydroceramide desaturase-1; FADS7; membrane fatty acid (lipid) desaturase; Membrane lipid desaturase; MGC5079; MIG15; migration-inducing gene 15 protein; MLD; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; Sphingolipid delta(4)-desaturase DES1
Gene Aliases: DEGS; DEGS-1; DEGS1; Des-1; DES1; FADS7; MIG15; MLD
UniProt ID: (Human) O15121
Entrez Gene ID: (Human) 8560
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.