Novus Biologicals
Manufacturer Code:NBP297548AF532
Catalog # NB044770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human DEFB118 fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta 18 beta-defensin 118 C20orf63 DEFB18 DEFB-18 defensin beta 118 ESC42 ESP13.6; Beta-defensin 118; Beta-defensin 18; DEFB-18; Defensin, beta 118; defensin, beta 18; Epididymal secretory protein 13.6; epididymus specific clone 42; ESP13.6
Gene Aliases: C20orf63; DEFB-18; DEFB118; DEFB18; ESC42; ESP13.6
UniProt ID: (Human) Q96PH6
Entrez Gene ID: (Human) 117285
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.