Proteintech
Manufacturer Code:147381AP150UL
Catalog # 50-172-9684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DEFB1 Fusion Protein Ag6406 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BD 1 BD1 Beta defensin 1 DEFB 1 DEFB1 DEFB101 defensin beta 1 hBD 1 HBD1; BD-1; beta defensin 1; Beta-defensin 1; beta-defensin-1; Defensin, beta 1
Gene Aliases: BD1; DEFB-1; DEFB1; DEFB101; HBD1
UniProt ID: (Human) P60022
Entrez Gene ID: (Human) 1672
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.