Abnova
Manufacturer Code:PAB25165
Catalog # 89-126-838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to DEFA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DEFA4.
For Research Use Only
Protein Aliases: corticostatin; DEF4; Defensin, alpha 4; defensin, alpha 4, corticostatin; defensin, alpha 4, preproprotein; HNP-4; HP-4; HP4; MGC120099; MGC138296; Neutrophil defensin 4
Gene Aliases: DEF4; DEFA4; HNP-4; HP-4; HP4
UniProt ID: (Human) P12838
Entrez Gene ID: (Human) 1669
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.