Invitrogen
Manufacturer Code:MA526116
Catalog # PIMA526116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G2 |
Immunogen | Full length human recombit protein of DEF6 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DEF-6; DEF6 guanine nucleotide exchange factor; DEF6 IBP differentially expressed in FDCP 6 homolog RP3-329A5.5 SLAT IRF4-binding protein OTTHUMP00000016251 differentially expressed in FDCP 6 homolog (mouse) SWAP70L SWAP-70-like adaptor protein of T cells; Differentially expressed in FDCP 6 homolog; IRF4-binding protein; SWAP-70-like adaptor protein of T cells
Gene Aliases: DEF6; IBP; SLAT; SWAP70L
UniProt ID: (Human) Q9H4E7
Entrez Gene ID: (Human) 50619, (Rhesus monkey) 718845, (Dog) 608163
Molecular Function:
calcium-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.