Invitrogen
Manufacturer Code:PA522193
Catalog # PIPA522193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 129 and 223 of DEDD (Uniprot IDNo.O75618) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDPro1; FLDED-1; KE05 CASP8IP1 DEDD1 DEFT FLDED1 DEDPro1 FLDED-1 death effector domain-containing protein death effector domain-containing testicular molecule
Gene Aliases: CASP8IP1; DEDD; DEDD1; DEDPRO1; DEFT; FLDED1; KE05
UniProt ID: (Human) O75618
Entrez Gene ID: (Human) 9191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.