Invitrogen
Manufacturer Code:PA5115110
Catalog # PIPA5115110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DEDD corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CASP8IP1 death effector domain containing death effector domain-containing death effector domain-containing protein Death effector domain-containing testicular molecule Dedd DEDD1 Dedpro1 Deft FLDED1 FLDED-1 KE05 tumor necrosis factor (ligand) superfamily member 19; Death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDPro1; FLDED-1; tumor necrosis factor (ligand) superfamily, member 19
Gene Aliases: CASP8IP1; DEDD; DEDD1; DEDPRO1; DEFT; FLDED1; KE05
UniProt ID: (Human) O75618, (Mouse) Q9Z1L3, (Rat) Q9Z2K0
Entrez Gene ID: (Human) 9191, (Mouse) 21945, (Rat) 83631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.