Invitrogen
Manufacturer Code:PA530503
Catalog # PIPA530503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 92 and 242 of Human DECR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,4-dienoyl CoA reductase 2, peroxisomal; 2,4-dienoyl-CoA reductase 2; pDCR; PDCR SDR17C1 24-dienoyl-CoA reductase 2 peroxisomal 24-dienoyl-CoA reductase short chain dehydrogenase/reductase family 17C member 1; Peroxisomal 2,4-dienoyl-CoA reductase [(3E)-enoyl-CoA-producing]; Short chain dehydrogenase/reductase family 17C member 1; short chain dehydrogenase/reductase family 17C, member 1
Gene Aliases: DECR2; PDCR; SDR17C1
UniProt ID: (Human) Q9NUI1
Entrez Gene ID: (Human) 26063
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.