Proteintech
Manufacturer Code:258551AP150UL
Catalog # 50-173-5102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DECR2 Fusion Protein Ag23052 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,4-dienoyl CoA reductase 2, peroxisomal; 2,4-dienoyl-CoA reductase 2; 24 dienoyl CoA reductase 2 DECR2 PDCR SDR17C1; pDCR; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal 2,4-dienoyl-CoA reductase [(3E)-enoyl-CoA-producing]; Short chain dehydrogenase/reductase family 17C member 1; short chain dehydrogenase/reductase family 17C, member 1
Gene Aliases: DECR2; PDCR; SDR17C1
UniProt ID: (Human) Q9NUI1, (Mouse) Q9WV68
Entrez Gene ID: (Human) 26063, (Mouse) 26378
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.