Novus Biologicals
Manufacturer Code:NBP272203C
Catalog # NBP272203C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-297 of human DEC2/SHARP1 (NP_110389) produced in E.coli. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: basic helix-loop-helix domain containing class B 3 basic helix-loop-helix family member e41 bHLHb3 BHLHB3class E basic helix-loop-helix protein 41 bHLHe41 Class B basic helix-loop-helix protein 3 DEC2Enhancer-of-split and hairy-related protein 1 Differentially expressed in chondrocytes protein 2 SHARP-1 SHARP1hDEC2; basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix family, member e41; bHLHb3; bHLHe41; Class B basic helix-loop-helix protein 3; Class E basic helix-loop-helix protein 41; Differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; SHARP-1
Gene Aliases: BHLHB3; BHLHE41; DEC2; hDEC2; SHARP1
UniProt ID: (Human) Q9C0J9
Entrez Gene ID: (Human) 79365
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.