Novus Biologicals
Manufacturer Code:NB1001800AF700
Catalog # NB1001800Z
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503] |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix domain containing class B 2 basic helix-loop-helix family member e40 bHLHb2 bHLHe40 Class B basic helix-loop-helix protein 2 class E basic helix-loop-helix protein 40 DEC1HLHB2 differentially expressed in chondrocytes 1 Differentially expressed in chondrocytes protein 1 differentiated embryo chondrocyte expressed gene 1 Enhancer-of-split and hairy-related protein 2 SHARP2 SHARP-2 Stimulated by retinoic acid gene 13 protein STRA13FLJ99214 Stra14; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix family, member e40; bHLHb2; bHLHe40; Class B basic helix-loop-helix protein 2; Class E basic helix-loop-helix protein 40; DEC1; differentially expressed in chondrocytes 1; Differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; Enhancer-of-split and hairy-related protein 2; SHARP-2; Stimulated by retinoic acid gene 13 protein
Gene Aliases: BHLHB2; BHLHE40; DEC1; HLHB2; SHARP-2; SHARP2; STRA13; Stra14
UniProt ID: (Human) O14503
Entrez Gene ID: (Human) 8553
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.