Invitrogen
Manufacturer Code:PA5112397
Catalog # PIPA5112397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Deformed epidermal autoregulatory factor 1 homolog protein (73-200AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU042387 C230009B13Rik Deaf1 DEAF-1 related transcriptional regulator (NUDR) DEAF1 transcription factor DEAF1 transcription factor deformed epidermal autoregulatory factor 1 (Drosophila) Deformed epidermal autoregulatory factor 1 homolog MRD24 Nuclear DEAF-1-related transcriptional regulator NUDR SPN Suppressin Zinc finger MYND domain-containing protein 5 ZMYND5; DEAF1 transcription factor; Deformed epidermal autoregulatory factor 1 homolog; Nuclear DEAF-1-related transcriptional regulator; NUDR; Suppressin; Zinc finger MYND domain-containing protein 5
Gene Aliases: DEAF1; MRD24; NUDR; SPN; ZMYND5
UniProt ID: (Human) O75398
Entrez Gene ID: (Human) 10522
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.