Invitrogen
Manufacturer Code:PA534442
Catalog # PIPA534442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid synthetic peptide near the center of human DDX60 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDX60 DEAD box protein 60 ATP-dependent RNA helicase DDX60; DEAD (Asp-Glu-Ala-Asp) box polypeptide 60; DEAD box protein 60; DEAD-box helicase 60; mCG11385-like; Probable ATP-dependent RNA helicase DDX60
Gene Aliases: 9830118M07; DDX60; RGD1560014
UniProt ID: (Human) Q8IY21
Entrez Gene ID: (Human) 55601, (Rat) 100360801, (Mouse) 234311
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.