Novus Biologicals
Manufacturer Code:NBP270563
Catalog # NBP270563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D6 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 1-298 of human DDX6 (NP_004388) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: ATP-dependent RNA helicase p54; ATP-dependent RNA helicase p54 DEAD (Asp-Glu-Ala-Asp) box polypeptide 6 DEAD box protein 6 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase 54kD) EC 3.6.1 EC 3.6.4.13 FLJ36338 HLR2DEAD box-6 Oncogene RCK probable ATP-dependent RNA helicase DDX6 RCKP54; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); Oncogene RCK; Probable ATP-dependent RNA helicase DDX6
Gene Aliases: DDX6; HLR2; P54; RCK
UniProt ID: (Human) P26196
Entrez Gene ID: (Human) 1656
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.