Novus Biologicals
Manufacturer Code:NB200192
Catalog # NB200192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 425 and the C-terminus (residue 483) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 6 using the numbering given Swiss-Prot entry P26196 (GeneID 1656). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase p54; ATP-dependent RNA helicase p54 DEAD (Asp-Glu-Ala-Asp) box polypeptide 6 DEAD box protein 6 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase 54kD) EC 3.6.1 EC 3.6.4.13 FLJ36338 HLR2DEAD box-6 Oncogene RCK probable ATP-dependent RNA helicase DDX6 RCKP54; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); Oncogene RCK; Probable ATP-dependent RNA helicase DDX6
Gene Aliases: DDX6; HLR2; P54; RCK
UniProt ID: (Human) P26196
Entrez Gene ID: (Human) 1656
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.