OriGene
Manufacturer Code:TA800438
Catalog # 50-167-8322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-303 of human DDX59 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; OFD5 ZNHIT5; Probable ATP-dependent RNA helicase DDX59; Zinc finger HIT domain-containing protein 5
Gene Aliases: DDX59; OFD5; ZNHIT5
UniProt ID: (Human) Q5T1V6
Entrez Gene ID: (Human) 83479
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.