OriGene
Manufacturer Code:TA506141S
Catalog # 50-167-6488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6C1 |
Immunogen | Full length human recombit protein of human DDX58 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antiviral innate immune response receptor RIG-I; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; Probable ATP-dependent RNA helicase DDX58; Retinoic acid-inducible gene 1 protein; Retinoic acid-inducible gene I protein; RIG-1; RIG-I; RIG-I RIGI RLR-1 SGMRT2; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I
Gene Aliases: DDX58; RIG-I; RIGI; RLR-1; SGMRT2
UniProt ID: (Human) O95786
Entrez Gene ID: (Human) 23586
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.