Novus Biologicals
Manufacturer Code:NBP246440
Catalog # NBP246440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E5 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 323-547 of human DDX56 (NP_061955) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 61-kd nucleolar helicase; ATP-dependent 61 kDa nucleolar RNA helicase; ATP-dependent 61 kDa nucleolar RNA helicase DDX26 DEAD (Asp-Glu-Ala-Asp) box polypeptide 56 DEAD box protein 21 DEAD box protein 56 DEAD-box RNA helicase EC 3.6.1 EC 3.6.4.1361-kd nucleolar helicase NOH61DDX21 probable ATP-dependent RNA helicase DDX56 putative nucleolar RNA helicase; DEAD (Asp-Glu-Ala-Asp) box helicase 56; DEAD (Asp-Glu-Ala-Asp) box polypeptide 56; DEAD box protein 21; DEAD box protein 56; DEAD-box RNA helicase; nucleolar helicase of 61 kDa; Probable ATP-dependent RNA helicase DDX56; putative nucleolar RNA helicase
Gene Aliases: DDX21; DDX26; DDX56; NOH61
UniProt ID: (Human) Q9NY93
Entrez Gene ID: (Human) 54606
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.