Invitrogen
Manufacturer Code:PA5116932
Catalog # PIPA5116932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human DDX55 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810021H22Rik ATP-dependent RNA helicase DDX55 DDX55 DEAD (Asp-Glu-Ala-Asp) box polypeptide 55 DEAD box protein 55 DEAD-box helicase 55 Kiaa1595 mKIAA1595; ATP-dependent RNA helicase DDX55; DEAD (Asp-Glu-Ala-Asp) box polypeptide 55; DEAD box protein 55
Gene Aliases: 2810021H22Rik; DDX55; KIAA1595; mKIAA1595
UniProt ID: (Human) Q8NHQ9, (Mouse) Q6ZPL9
Entrez Gene ID: (Human) 57696, (Mouse) 67848
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.