Novus Biologicals
Manufacturer Code:NBP15733120UL
Catalog # NBP1573320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DDX55 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 55) The peptide sequence was selected from the N terminal of DDX55. Peptide sequence RELAIQIDEVLSHFTKHFPEFSQILWIGGRNPGEDVERFKQQGGNIIVAT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX55; DEAD (Asp-Glu-Ala-Asp) box polypeptide 55; DEAD (Asp-Glu-Ala-Asp) box polypeptide 55 DEAD box protein 55 EC 3.6.1 EC 3.6.4.13 FLJ16577 KIAA1595ATP-dependent RNA helicase DDX55 MGC33209; DEAD box protein 55
Gene Aliases: DDX55; KIAA1595
UniProt ID: (Human) Q8NHQ9
Entrez Gene ID: (Human) 57696
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.