R&D Systems
Manufacturer Code:MAB6370SP
Catalog # MAB6370SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 632813 |
Immunogen | <EM>E. coli</EM>-derived recombinant human DDX5 Asn448-Gln614 Accession # P17844 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX5; B-cell CLL/lymphoma 7 protein family member C B-cell CLL/lymphoma 7C DEAD/H G17P1 HELR HLR1 HUMP68; DEAD (Asp-Glu-Ala-Asp) box helicase 5; DEAD (Asp-Glu-Ala-Asp) box polypeptide 5; DEAD box protein 5; DEAD box-5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase, 68kD); Probable ATP-dependent RNA helicase DDX5; RNA helicase p68
Gene Aliases: DDX5; G17P1; HELR; HLR1; HUMP68; p68
UniProt ID: (Human) P17844
Entrez Gene ID: (Human) 1655
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.