Invitrogen
Manufacturer Code:PA527131
Catalog # PIPA527131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 552 and 614 of Human DDX5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX5; DEAD (Asp-Glu-Ala-Asp) box helicase 5; DEAD (Asp-Glu-Ala-Asp) box polypeptide 5; DEAD box protein 5; DEAD box-5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase, 68kD); G17P1 HLR1 HUMP68 p68 ATP-dependent RNA helicase DDX5 DEAD box protein 5 DEAD box-5 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase 68kD) RNA helicase p68 probable ATP-dependent RNA helicase DDX5; Probable ATP-dependent RNA helicase DDX5; RNA helicase p68
Gene Aliases: DDX5; G17P1; HELR; HLR1; HUMP68; p68
UniProt ID: (Human) P17844
Entrez Gene ID: (Human) 1655
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.