Invitrogen
Manufacturer Code:PA518260
Catalog # PIPA518260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PMIGYPMPTGYSQ) corresponding to the C-terminus amino acids of DDX5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX5; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 5; DDX5 P68 HLR1 G17P1 HUMP68 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase 68kDa) RNA-dependent ATPase DEAD/H box-5 (RNA helicase 68kD) DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase 68kD) DEAD (Asp-Glu-Ala-Asp) box polypeptide 5 DKFZp434E109 DKFZp686J01190 ATP-dependent RNA helicase DDX5 DEAD box-5; DEAD (Asp-Glu-Ala-Asp) box helicase 5; DEAD (Asp-Glu-Ala-Asp) box polypeptide 5; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 5; DEAD box protein 5; DEAD box RNA helicase DEAD1; DEAD box-5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase, 68kD); mDEAD1; p68 RNA helicase; Probable ATP-dependent RNA helicase DDX5; RNA helicase p68
Gene Aliases: 2600009A06Rik; DDX5; G17P1; HELR; HLR1; HUMP68; p68
UniProt ID: (Human) P17844
Entrez Gene ID: (Human) 1655, (Mouse) 13207
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.