Abnova
Manufacturer Code:H00009775A01
Catalog # 89-106-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 312 a.a. to 411 a.a) partial recombinant protein with GST tag.,DDX48 (NP_055555 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant DDX48.
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-3; DDX48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; eIF-4A-III; eIF4A-III; eIF4AIII; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-like NUK-34; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 3; hNMP 265; hNMP265; KIAA0111; MGC10862; NMP 265; NMP265; Nuclear matrix protein 265; NUK-34
Gene Aliases: DDX48; EIF4A3; eIF4AIII; KIAA0111; MUK34; NMP265; NUK34; RCPS
UniProt ID: (Human) P38919
Entrez Gene ID: (Human) 9775
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.