Novus Biologicals
Manufacturer Code:NBP187134
Catalog # NBP187134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FGATSSSSGFGKSAPPQLPSFYKIGSKRANFDEENAYFEDEEEDSSNVDLPYIPAENSPTRQQFHSKPVDSDSDDDPLEAFMAEVEDQAARDMKRLEEKDKERKNVKGIRDD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX42; DEAD (Asp-Glu-Ala-Asp) box helicase 42; DEAD (Asp-Glu-Ala-Asp) box polypeptide 42; DEAD (Asp-Glu-Ala-Asp) box polypeptide 42 DEAD box protein 42 EC 3.6.1 EC 3.6.4.13 RHELPSF3b DEAD box protein RNA helicase-like protein RNA helicase-related protein RNAHPFLJ43179 SF3b125 DEAD-box protein SF3b125ATP-dependent RNA helicase DDX42 Splicing factor 3B-associated 125 kDa protein; DEAD box protein 42; RHELP; RNA helicase-like protein; RNA helicase-related protein; RNAHP; SF3b DEAD box protein; SF3b125; splicing factor 3b, subunit 8; Splicing factor 3B-associated 125 kDa protein
Gene Aliases: DDX42; DDX42P; RHELP; RNAHP; SF3b125; SF3B8
UniProt ID: (Human) A8KA43
Entrez Gene ID: (Human) 11325
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.