Invitrogen
Manufacturer Code:PA583928
Catalog # PIPA583928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DDX41 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900024F02Rik; ABS MGC8828; DEAD (Asp-Glu-Ala-Asp) box polypeptide 41; DEAD box protein 41; DEAD box protein abstrakt homolog; DEAD-box protein abstrakt; Probable ATP-dependent RNA helicase DDX41; putative RNA helicase
Gene Aliases: ABS; DDX41; MPLPF
UniProt ID: (Human) Q9UJV9
Entrez Gene ID: (Human) 51428
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.