Abnova
Manufacturer Code:H00051428A01
Catalog # 89-106-899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 523 a.a. to 622 a.a) partial recombinant protein with GST tag.,DDX41 (NP_057306 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant DDX41.
For Research Use Only
Protein Aliases: 2900024F02Rik; ABS; DEAD (Asp-Glu-Ala-Asp) box polypeptide 41; DEAD box protein 41; DEAD box protein abstrakt homolog; DEAD-box protein abstrakt; MGC8828; Probable ATP-dependent RNA helicase DDX41; putative RNA helicase
Gene Aliases: ABS; DDX41; MPLPF
UniProt ID: (Human) Q9UJV9
Entrez Gene ID: (Human) 51428
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.