Invitrogen
Manufacturer Code:PA522050
Catalog # PIPA522050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 70 and 346 of DDX3 (Uniprot IDNo.O15523) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX3Y; D1Pas1 related sequence 1; D1Pas1-related sequence 1; DBY ATP-dependent RNA helicase DDX3Y DEAD box protein 3 Y-chromosomal DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide Y chromosome; DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide, Y chromosome; DEAD box protein 3, Y-chromosomal; DEAD-box RNA helicase DEAD2; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, Y chromosome; mDEAD2; uncharacterized protein LOC364073
Gene Aliases: 8030469F12Rik; D1Pas1-rs1; DBY; DDX3Y; Dead2; RGD1309586
UniProt ID: (Human) O15523, (Mouse) Q62095
Entrez Gene ID: (Human) 8653, (Rat) 364073, (Mouse) 26900
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.