OriGene
Manufacturer Code:TA812666
Catalog # 50-168-4642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 319-428 of human DDX39B produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 56 kDa U2AF65-associated protein; ATP-dependent RNA helicase p47; BAT1 D6S81E UAP56; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B; DEAD box protein UAP56; DEAD-box helicase 39B; HLA-B associated transcript 1; HLA-B-associated transcript 1 protein; HLA-B-associated transcript 1A; nuclear RNA helicase (DEAD family); nuclear RNA helicase Bat1; spliceosome RNA helicase BAT1; Spliceosome RNA helicase DDX39B
Gene Aliases: 0610030D10Rik; AI428441; Bat-1; BAT1; Bat1a; D17H6S81E; D17H6S81E-1; D6S81E; D6S81Eh; DDX39B; UAP56
UniProt ID: (Human) Q13838, (Dog) Q5WR10, (Mouse) Q9Z1N5
Entrez Gene ID: (Human) 7919, (Dog) 474839, (Mouse) 53817
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.