Invitrogen
Manufacturer Code:PA5109624
Catalog # PIPA5109624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 178-427 of human DDX39A (NP_005795.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610307C23Rik ATP-dependent RNA helicase DDX39A BAT1 BAT1L DDX39 Ddx39a Ddxl DEAD box protein 39 Nuclear RNA helicase URH49 Nuclear RNA helicase DECD variant of DEAD box family URH49; ATP-dependent RNA helicase DDX39; ATP-dependent RNA helicase DDX39A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; Nuclear RNA helicase URH49; Nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa
Gene Aliases: 2610307C23Rik; BAT1; BAT1L; DDX39; DDX39A; DDXL; URH49
UniProt ID: (Human) O00148, (Mouse) Q8VDW0, (Rat) Q5U216
Entrez Gene ID: (Human) 10212, (Mouse) 68278, (Rat) 89827
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.