Abnova
Manufacturer Code:MAB6576
Catalog # 89-112-943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E4 |
Immunogen | A synthetic peptide corresponding to N-terminus of human DDX39. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of DDX39.
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX39A; BAT1; BAT1L; DDXL; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; MGC18203; MGC8417; Nuclear RNA helicase URH49; nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa; URH49
Gene Aliases: BAT1; BAT1L; DDX39; DDX39A; DDXL; URH49
UniProt ID: (Human) O00148
Entrez Gene ID: (Human) 10212
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.