Novus Biologicals
Manufacturer Code:NBP292080100UL
Catalog # NB026052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 178-427 of human DDX39A (NP_005795.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX39 BAT1 BAT1L DDX39 DDXL DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 transcript DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A DEAD box protein 39 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39 EC 3.6.4.13 MGC18203 MGC8417 Nuclear RNA helicase URH49 nuclear RNA helicase DECD variant of DEAD box family UAP56-related helicase 49 kDa URH49; ATP-dependent RNA helicase DDX39A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; Nuclear RNA helicase URH49; nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa
Gene Aliases: BAT1; BAT1L; DDX39; DDX39A; DDXL; URH49
UniProt ID: (Human) O00148
Entrez Gene ID: (Human) 10212
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.