Invitrogen
Manufacturer Code:PA590797
Catalog # PIPA590797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 652-851 of human DDX31 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DEAD (Asp-Glu-Ala-Asp) box polypeptide 31; DEAD box protein 31; DEAD/DEXH helicase DDX31; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 31; G2 helicase; Helicain; Probable ATP-dependent RNA helicase DDX31; protein phosphatase 1, regulatory subunit 25
Gene Aliases: DDX31; PPP1R25
UniProt ID: (Human) Q9H8H2
Entrez Gene ID: (Human) 64794
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.