Novus Biologicals
Manufacturer Code:NBP132780
Catalog # NBP132780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human DDX3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX3X; CAP-Rf; DBX; DBXCAP-Rf DDX14 DDX3DEAD/H box-3 DEAD (Asp-Glu-Ala-Asp) box polypeptide 3 X-linked DEAD box protein 3 X-chromosomal DEAD box X isoform DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3 EC 3.6.1 EC 3.6.4.13 helicase like protein 2 Helicase-like protein 2 HLP2ATP-dependent RNA helicase DDX3X; DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked; DEAD box protein 3, X-chromosomal; DEAD box, X isoform; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3; DEAD/H box-3; Helicase-like protein 2; HLP2
Gene Aliases: CAP-Rf; DBX; DDX14; DDX3; DDX3X; HLP2; MRX102
UniProt ID: (Human) O00571
Entrez Gene ID: (Human) 1654
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.