Novus Biologicals
Manufacturer Code:NBP292376100UL
Catalog # NB026633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 371-540 of human DDX28 (NP_060850.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DEAD (Asp-Glu-Ala-Asp) box polypeptide 28; DEAD (Asp-Glu-Ala-Asp) box polypeptide 28 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28 EC 3.6.1 EC 3.6.4.13 FLJ11282 M MDDX28probable ATP-dependent RNA helicase DDX28 Mitochondrial DEAD box protein 28 mitochondrial DEAD-box polypeptide 28; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28; Mitochondrial DEAD box protein 28; mitochondrial DEAD-box polypeptide 28; Probable ATP-dependent RNA helicase DDX28
Gene Aliases: DDX28; MDDX28
UniProt ID: (Human) Q9NUL7
Entrez Gene ID: (Human) 55794
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.