Invitrogen
Manufacturer Code:MA536115
Catalog # PIMA536115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A3B10 |
Immunogen | Recombinant protein within zebrafish ddx24 aa 550-820. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1700055J08Rik 2510027P10Rik AI649272 ATP-dependent RNA helicase DDX24 Ddx24 DEAD (Asp-Glu-Ala-Asp) box helicase 24 DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 DEAD box protein 24 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A) DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24 DEAD-box helicase 24 S. cerevisiae CHL1-like helicase; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24
Gene Aliases: si:ch1073-416d2.2
Entrez Gene ID: (Zebrafish) 553462
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.