Proteintech
Manufacturer Code:157691AP150UL
Catalog # 50-173-0463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DDX24 Fusion Protein Ag8407 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX24; DDX24 DEAD box protein 24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; S. cerevisiae CHL1-like helicase
Gene Aliases: 1700055J08Rik; 2510027P10Rik; AI649272; DDX24
UniProt ID: (Human) Q9GZR7, (Mouse) Q9ESV0
Entrez Gene ID: (Human) 57062, (Rat) 373065, (Mouse) 27225
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.