Proteintech
Manufacturer Code:101992AP150UL
Catalog # 50-172-6104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DDX23 Fusion Protein Ag0258 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa U5 snRNP-specific protein; DDX23 DEAD box protein 23 prp28 PRP28 homolog PRPF28 U5 100K U5 100KD; DEAD (Asp-Glu-Ala-Asp) box polypeptide 23; DEAD box protein 23; Probable ATP-dependent RNA helicase DDX23; PRP28 homolog; PRP28 homolog, yeast; PRP28p homolog; U5 snRNP 100 kD protein; U5-100kD
Gene Aliases: 3110082M05Rik; 4921506D17Rik; DDX23; prp28; PRPF28; SNRNP100; U5-100K; U5-100KD
UniProt ID: (Human) Q9BUQ8
Entrez Gene ID: (Human) 9416, (Mouse) 74351, (Rat) 300208
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.