Abnova
Manufacturer Code:H00009416K
Catalog # 89-206-353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human DDX23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human DDX23 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 100 kDa U5 snRNP-specific protein; DEAD (Asp-Glu-Ala-Asp) box polypeptide 23; DEAD box protein 23; MGC8416 PRPF28 U5-100K U5-100KD prp28; Probable ATP-dependent RNA helicase DDX23; PRP28 homolog; PRP28 homolog, yeast; PRP28p homolog; U5 snRNP 100 kD protein; U5-100kD
Gene Aliases: DDX23; prp28; PRPF28; SNRNP100; U5-100K; U5-100KD
UniProt ID: (Human) Q9BUQ8
Entrez Gene ID: (Human) 9416
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.