Proteintech
Manufacturer Code:113241AP150UL
Catalog # 50-172-7061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DDX20 Fusion Protein Ag1863 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Component of gems 3; Component of gems 3 DDX20 DEAD box protein 20 DEAD box protein DP 103 DP103 Gemin 3 GEMIN3; DEAD (Asp-Glu-Ala-Asp) box polypeptide 20; DEAD box protein 20; DEAD box protein DP 103; DEAD-box protein DP103; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD; Gemin-3; Probable ATP-dependent RNA helicase DDX20; Regulator of steroidogenic factor 1; ROSF-1; SMN-interacting protein
Gene Aliases: DDX20; DP103; GEMIN3
UniProt ID: (Human) Q9UHI6, (Mouse) Q9JJY4
Entrez Gene ID: (Human) 11218, (Rat) 84473, (Mouse) 53975
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.