Invitrogen
Manufacturer Code:PA541737
Catalog # PIPA541737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DDX19B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX19; ATP-dependent RNA helicase DDX19B; DEAD (Asp-Glu-Ala-As) box polypeptide 19B; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B DBP5 RNAh DDX19; DEAD box protein 19B; DEAD box RNA helicase DEAD5; DEAD-box protein 5; DEAD-box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5, yeast, homolog); yeast Dbp5 homolog; zinc responsive protein Zd10A
Gene Aliases: 2810457M08Rik; 4921519L13Rik; AW260119; DBP5; DDX19; DDX19B; RNAh; TDBP; Zd10a
UniProt ID: (Human) Q9UMR2
Entrez Gene ID: (Dog) 479657, (Human) 11269, (Pig) 100524746, (Bovine) 517438, (Guinea pig) 100719332, (Mouse) 234733, (Rat) 690693
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.