Novus Biologicals
Manufacturer Code:NBP216126
Catalog # NBP216126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human DDX19B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX19; ATP-dependent RNA helicase DDX19 DBP5ATP-dependent RNA helicase DDX19B DDX19DEAD-box protein 5 DEAD (Asp-Glu-Ala-As) box polypeptide 19 DEAD (Asp-Glu-Ala-As) box polypeptide 19B DEAD box protein 19B DEAD box RNA helicase DEAD5 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5 yeast homolog) DEAD-box RNA helicase DEAD5 EC 3.6.1 EC 3.6.4.13 RNAh yeast Dbp5 homolog; ATP-dependent RNA helicase DDX19B; DEAD (Asp-Glu-Ala-As) box polypeptide 19B; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B; DEAD box protein 19B; DEAD box RNA helicase DEAD5; DEAD-box protein 5; DEAD-box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5, yeast, homolog); yeast Dbp5 homolog
Gene Aliases: DBP5; DDX19; DDX19B; RNAh; TDBP
UniProt ID: (Human) Q9UMR2
Entrez Gene ID: (Human) 11269
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.