Novus Biologicals
Manufacturer Code:NBP297530JF549
Catalog # NB044587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human DDX19A fragment. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX19A; ATP-dependent RNA helicase DDX19A DDX19-DDX19L DDX19L DDX19-like protein DEAD (Asp-Glu-Ala-As) box polypeptide 19A DEAD (Asp-Glu-Ala-As) box polypeptide 19-like DEAD box protein 19A DKFZp686C21137 EC 3.6.1 EC 3.6.4.13 FLJ11126 RNA helicase; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19A; DEAD box protein 19A; RNA helicase
Gene Aliases: DDX19-DDX19L; DDX19A; DDX19L
UniProt ID: (Human) Q9NUU7
Entrez Gene ID: (Human) 55308
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.