R&D Systems
Manufacturer Code:AF4530
Catalog # AF4530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DDX17 Asn498-Tyr638 Accession # Q92841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DEAD (Asp-Glu-Ala-Asp) box helicase 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 DEAD box protein 17 DEAD box protein p72 DEAD/H box 17 DKFZp761H2016 EC 3.6.1 EC 3.6.4.13 p72 P72DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD) probable ATP-dependent RNA helicase DDX17 RH70 RNA-dependent helicase p72; DEAD box protein 17; DEAD box protein p72; DEAD box protein p82; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD); Probable ATP-dependent RNA helicase DDX17; RNA-dependent helicase p72
Gene Aliases: DDX17; P72; RH70
UniProt ID: (Human) Q92841
Entrez Gene ID: (Human) 10521
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.