Invitrogen
Manufacturer Code:PA583227
Catalog # PIPA583227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DDX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX1; DBP-RB; DEAD (Asp-Glu-Ala-Asp) box helicase 1; DEAD (Asp-Glu-Ala-Asp) box polypeptide 1; DEAD box polypeptide 1; DEAD box protein 1; DEAD box protein retinoblastoma; DEAD box-1; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1
Gene Aliases: AA409185; DBP-RB; DDX1; UKVH5d
UniProt ID: (Human) Q92499, (Rat) Q641Y8, (Mouse) Q91VR5
Entrez Gene ID: (Human) 1653, (Rat) 84474, (Mouse) 104721
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.