Invitrogen
Manufacturer Code:PA521386
Catalog # PIPA521386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 118 of DDT (Uniprot IDNo.P30046) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-dopachrome decarboxylase; D-dopachrome tautomerase; hCG&lowbar41098 DDCT D-dopachrome decarboxylase phenylpyruvate tautomerase II; Phenylpyruvate tautomerase II
Gene Aliases: DDCT; DDT
UniProt ID: (Human) P30046
Entrez Gene ID: (Human) 1652
Molecular Function: cytokine isomerase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.